5-Acetylsalicylamide

Supplier: Thermo Scientific

SB01472.1 SB01472.10 SB01472EE
MAYBSB01472.1EA 0 EUR
MAYBSB01472.1 MAYBSB01472.10 MAYBSB01472EE
5-Acetylsalicylamide
5-Acetylsalicylamide
Formula: C₉H₉NO₃
MDL Number: MFCD00049222
CAS Number: 40187-51-7
EINECS: 254-830-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR