Nordihydroguaiaretic acid 95%
Supplier: Thermo Fisher Scientific
Warning
376300010 376300050 376305000
ACRO376300010EA 80 EUR
ACRO376300010 ACRO376300050 ACRO376305000
Nordihydroguaiaretic acid 95%
Nordihydroguaiaretic acid
Formula: C₁₈H₂₂O₄
MW: 302.37 g/mol
Melting Pt: 184…189 °C
Storage Temperature: Freezer
MDL Number: MFCD00002206
CAS Number: 500-38-9
EINECS: 207-903-0
Order Now
Specification Test Results
Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Melting point 184 to 189 °C
HPLC ≥94 %