Sodium phenoxide trihydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium phenolate trihydrate
MQ100
318191-100G 318191-5G
SIAL318191-100GEA 60.3 EUR
SIAL318191-100G SIAL318191-5G
Sodium phenoxide trihydrate, Sigma-Aldrich®
Sodium phenoxide trihydrate
Formula: C₆H₅NaO·3H₂O
MDL Number: MFCD00149636
CAS Number: 156150-40-2
EINECS: 205-347-3