Nickel(II) wolframate ≥99.9% (metals basis excluding Co) (max. 100 ppm Co)
Supplier: Thermo Scientific
Danger
Synonyms: Nickel tungsten oxide
41826.14 41826.22
41826.14EA 183 EUR
41826.14 41826.22
Nickel(II) wolframate ≥99.9% (metals basis excluding Co) (max. 100 ppm Co)
Nickel(II) wolframate
Purity: 99.9% CAS No.: 14177-51-6 Molecular Formula: NiWO4 Formula Weight: 306.53 Physical Form: -100 Mesh Powder Odour: Odourless MDL No.: MFCD00016268
Formula: NiO₄W
MW: 306.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016268
CAS Number: 14177-51-6
EINECS: 238-032-4