4-Acetyl-2-chloropyridine ≥97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1-(2-chloropyridin-4-yl)ethan-1-one, 1-(2-chloropyridine-4-yl)ethanone
H33619.03 H33619.06
H33619.03EA 102 EUR
H33619.03 H33619.06
4-Acetyl-2-chloropyridine ≥97%
4-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.58 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD07699406
CAS Number: 23794-15-2
UN: 2811
ADR: 6.1 III