5-(4-Morpholinyl)-2-nitrophenol ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B24950.03
B24950.06
B24950.03EA
34.3
EUR
B24950.03
B24950.06
5-(4-Morpholinyl)-2-nitrophenol ≥97%
5-(4-Morpholinyl)-2-nitrophenol
Formula:
C₁₀H₁₂N₂O₄ Melting Pt: 143…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053057 CAS Number: 175135-19-0 |