10-Bromodecanoic acid 95%
Supplier: Thermo Fisher Scientific
Warning
H60292.06 H60292.14
H60292.06EA 57.1 EUR
H60292.06 H60292.14
10-Bromodecanoic acid 95%
10-Bromodecanoic acid
Formula: C₁₀H₁₉BrO₂
Melting Pt: 38…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00014388
CAS Number: 50530-12-6