10-Bromodecanoic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

541397-25G 541397-5G
SIAL541397-25GEA 48.4 EUR
SIAL541397-25G SIAL541397-5G
10-Bromodecanoic acid, Sigma-Aldrich®
10-Bromodecanoic acid
Formula: C₁₀H₁₉BrO₂
MW: 251.16 g/mol
MDL Number: MFCD00014388
CAS Number: 50530-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR