Zinc chloride 1.9 mol/l, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Zinc dichloride, Dichlorozinc, Zinc (II) chloride
MQ100
703516-100ML
703516-800ML
SIAL703516-100MLEA
172
EUR
SIAL703516-100ML
SIAL703516-800ML
Zinc chloride 1.9 mol/l, Sigma-Aldrich®
Zinc chloride
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 Melting Pt: 283 |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 Merck Index: 13,10185 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...