10-Hydroxydecanoic acid 96%
Supplier: Thermo Scientific
H60743.03 H60743.06
H60743.03EA 120 EUR
H60743.03 H60743.06
10-Hydroxydecanoic acid 96%
10-Hydroxydecanoic acid
Formula: C₁₀H₂₀O₃
MW: 188.26 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00010510
CAS Number: 1679-53-4
EINECS: 216-848-1