Tin(IV) tert-butanolate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Tin(IV) tert-butoxide, Tin(IV) t-butoxide
M-Clarity™ quality level = MQ100
494135-25G
494135-5G
SIAL494135-25GEA
548
EUR
SIAL494135-25G
SIAL494135-5G
Tin(IV) tert-butanolate, Sigma-Aldrich®
Tin (IV) tert-butanolate
Formula:
C₁₆H₃₆O₄Sn MW: 411.165 g/mol |
MDL Number:
MFCD00156551 CAS Number: 36809-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...