Potassium stannate trihydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Potassium tin(IV) oxide trihydrate
M-Clarity™ quality level = MQ100
462799-1KG 462799-250G
SIAL462799-1KGEA 138 EUR
SIAL462799-1KG SIAL462799-250G
Potassium stannate trihydrate, Sigma-Aldrich®
Potassium stannate trihydrate
Formula: K₂O₃Sn·3H₂O
MW: 298.94 g/mol (244.89 anhy)
MDL Number: MFCD00150391
CAS Number: 12125-03-0
EINECS: 235-255-9