6-Bromobenzimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms:
6-Bromo-1H-benzimidazole
702188-1G
702188-5G
SIAL702188-1GEA
33.8
EUR
SIAL702188-1G
SIAL702188-5G
6-Bromobenzimidazole, Sigma-Aldrich®
5-Bromobenzimidazole
Formula:
C₇H₅BrN₂ MW: 197.04 g/mol |
MDL Number:
MFCD00460014 CAS Number: 4887-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...