1,3-Dimethylimidazolium dimethylphosphate ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: 1,3-Dimethylimidazolium Dimethyl Phosphate
H27788.18 H27788.06
H27788.18EA 175 EUR
H27788.18 H27788.06
1,3-Dimethylimidazolium dimethylphosphate ≥98%
1,3-Dimethylimidazolium dimethylphosphate
Purity: 98% CAS No.: 654058-04-5 Molecular Formula: C7H15N2O4P Formula Weight: 222.18 MDL No.: MFCD08275369
Formula: C₇H₁₅N₂O₄P
MW: 222.18 g/mol
Density: 1.277
Storage Temperature: Ambient
MDL Number: MFCD08275369
CAS Number: 654058-04-5