meso-1,2-Dibromo-1,2-diphenylethane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
106550-100G 106550-25G
SIAL106550-100GEA 256 EUR
SIAL106550-100G SIAL106550-25G
meso-1,2-Dibromo-1,2-diphenylethane, Sigma-Aldrich®
meso-1,2-Dibromo-1,2-diphenylethane
Formula: C₁₄H₁₂Br₂
MW: 340.07 g/mol
MDL Number: MFCD00000137
CAS Number: 13440-24-9