1-Methoxy-1,3-butadiene (mixture of cis and trans isomeres), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
M12002-1G M12002-5G
SIALM12002-1GEA 164 EUR
SIALM12002-1G SIALM12002-5G
1-Methoxy-1,3-butadiene (mixture of cis and trans isomeres), Sigma-Aldrich®
1-Methoxy-1,3-butadiene (mixture of cis and trans isomeres)
Formula: C₅H₈O
MDL Number: MFCD00008660
CAS Number: 3036-66-6
EINECS: 221-234-1