1-(4-Bromophenyl)piperidine 97
Supplier: Thermo Fisher Scientific
Warning
H57102.03 H57102.06
H57102.03EA 83.3 EUR
H57102.03 H57102.06
1-(4-Bromophenyl)piperidine 97
1-(4-Bromophenyl)piperidine
Purity: 97% CAS No.: 22148-20-5 Molecular Formula: C11H14BrN Formula Weight: 240.14 MDL No.: MFCD04112533
Formula: C₁₁H₁₄BrN
MW: 240.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112533
CAS Number: 22148-20-5