6,6'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6,6'-Dimethyl-2,2'-bipyridine

M-Clarity™ quality level = MQ100

496367-5G 496367-1G
SIAL496367-5GEA 254 EUR
SIAL496367-5G SIAL496367-1G
6,6'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®
6,6'-Dimethyl-2,2'-bipyridyl
Formula: C₁₂H₁₂N₂
MW: 184.2371 g/mol
MDL Number: MFCD00059776
CAS Number: 4411-80-7
EINECS: 224-566-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR