Ethyl-3-piperidinecarboxylate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethyl nipecotate, Ethyl 3-Piperidinecarboxylate, Ethyl piperidine-3-carboxylate

M-Clarity™ quality level = MQ100

194360-100G 194360-25G
SIAL194360-100GEA 89.1 EUR
SIAL194360-100G SIAL194360-25G
Ethyl-3-piperidinecarboxylate, Sigma-Aldrich®
Ethyl 3-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD00005991
CAS Number: 5006-62-2
EINECS: 225-681-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR