2-Bromo-6-fluorobenzoic acid ≥97%
Supplier: Thermo Fisher Scientific
Warning
H61251.03 H61251.06
H61251.03EA 60.4 EUR
H61251.03 H61251.06
2-Bromo-6-fluorobenzoic acid ≥97%
2-Bromo-6-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD01569539
CAS Number: 2252-37-1