6-Methyl-2-pyridone
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-6-methylpyridine, 6-Methylpyridin-2-ol, 6-Methylpyridin-2(1H)-one
SB01977.1
SB01977EA
SB01977EE
MAYBSB01977.1EA
0
EUR
MAYBSB01977.1
MAYBSB01977EA
MAYBSB01977EE
6-Methyl-2-pyridone
6-Methyl-2-pyridone
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 283…284 °C (1013 hPa) Melting Pt: 159…160 °C Density: 1,13 g/cm³ (20 °C) |
MDL Number:
MFCD00006279 CAS Number: 3279-76-3 EINECS: 221-919-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...