ω-Bromo-2,4-dichloroacetophenone 95%
Supplier: Thermo Scientific
Danger
Synonyms: 2-Bromo-1-(2,4-dichlorophenyl)ethan-1-one, 2,4-Dichlorophenacyl bromide, α-Bromo-2,4-dichloroacetophenone, 2-Bromo-2',4'-dichloroacetophenone, 2-Bromo-1-(2,4-dichlorophenyl)ethanone
H59615.03 H59615.06
H59615.03EA 76.3 EUR
H59615.03 H59615.06
ω-Bromo-2,4-dichloroacetophenone 95%
ω-Bromo-2,4-dichloroacetophenone
Formula: C₈H₅BrCl₂O
MW: 267.94 g/mol
Melting Pt: 25…29 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053005
CAS Number: 2631-72-3
EINECS: 220-116-7
UN: 1759
ADR: 8,III