1-Pentyl-4-ethynylbenzene ≥97%
Supplier: Thermo Fisher Scientific
H60913.09 H60913.04
H60913.09EA 153 EUR
H60913.09 H60913.04
1-Pentyl-4-ethynylbenzene ≥97%
1-Pentyl-4-ethynylbenzene
Formula: C₁₃H₁₆
Boiling Pt: 172 °C
Density: 0.885
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00173884
CAS Number: 79887-10-8