2-Phenylthiophene 95%
Supplier: Thermo Scientific
H60819.03 H60819.06
H60819.03EA 46 EUR
H60819.03 H60819.06
2-Phenylthiophene 95%
2-Phenylthiophene
Formula: C₁₀H₈S
MW: 160.24 g/mol
Boiling Pt: 256 °C
Melting Pt: 34…36 °C
Flash Pt: 109 °C (228 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00130080
CAS Number: 825-55-8