4-Fluoro-o-phenylenediamine

Supplier: Thermo Scientific

Synonyms: 3,4-Diamino-1-fluorobenzene, 4-Fluorobenzene-1,2-diamine, 1,2-Diamino-4-fluorobenzene, 4-Fluoro-1,2-phenylenediamine

BTB02218.1 BTB02218.10 BTB02218.50
MAYBBTB02218.1EA 0 EUR
MAYBBTB02218.1 MAYBBTB02218.10 MAYBBTB02218.50
4-Fluoro-o-phenylenediamine
4-Fluoro-o-phenylenediamine
Formula: C₆H₇FN₂
MW: 126.13 g/mol
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 92 °C
MDL Number: MFCD00042228
CAS Number: 367-31-7
EINECS: 206-691-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR