5-Methoxy-o-toluidine ≥97%
Supplier: Thermo Scientific
Danger
Synonyms: 2-Amino-4-methoxytoluene, 3-Amino-4-methylanisole, 1-Amino-3-methoxy-6-methylbenzene, 6-Methyl-m-anisidine, 5-Methoxy-2-methylaniline
H60177.03 H60177.06
H60177.03EA 47.4 EUR
H60177.03 H60177.06
5-Methoxy-o-toluidine ≥97%
5-Methoxy-o-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 43…46 °C
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075057
CAS Number: 50868-72-9
EINECS: 256-816-4
UN: 2811
ADR: 6.1,III