1-Ethyl-3-methylimidazolium methylsulphate ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium methyl sulphate
H59480.06 H59480.18
H59480.06EA 39.9 EUR
H59480.06 H59480.18
1-Ethyl-3-methylimidazolium methylsulphate ≥98%
1-Ethyl-3-methylimidazolium methyl sulphate
Formula: C₇H₁₄N₂O₄S
MW: 222.27 g/mol
Density: 1.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD08457697
CAS Number: 516474-01-4