1-Bromo-2-fluoro-4-nitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Bromo-3-fluoronitrobenzene
H61494.14 H61494.22
H61494.14EA 128 EUR
H61494.14 H61494.22
1-Bromo-2-fluoro-4-nitrobenzene ≥98%
1-Bromo-2-fluoro-4-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 88…94 °C
Storage Temperature: Ambient
MDL Number: MFCD05865088
CAS Number: 185331-69-5