1-Bromo-5-fluoro-2-nitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Bromo-4-fluoro-1-nitrobenzene, 2-Bromo-4-fluoronitrobenzene
H61466.06 H61466.14
H61466.06EA 59.4 EUR
H61466.06 H61466.14
1-Bromo-5-fluoro-2-nitrobenzene ≥98%
1-Bromo-5-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 41…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00792441
CAS Number: 700-36-7