1-Methyl-3-octylimidazolium chloride 97
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-Methyl-3-n-octylimidazolium chloride
H59534.06 H59534.18
H59534.06EA 36 EUR
H59534.06 H59534.18
1-Methyl-3-octylimidazolium chloride 97
1-Methyl-3-octylimidazolium chloride
Formula: C₁₂H₂₃ClN₂
MW: 230.78 g/mol
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD03095432
CAS Number: 64697-40-1