(2-Fluorophenyl)acetone ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Fluorophenylacetone, 1-(2-Fluorophenyl)propan-2-one, 1-(2-Fluorophenyl)-2-propanone
H61535.06 H61535.14
H61535.06EA 68 EUR
H61535.06 H61535.14
(2-Fluorophenyl)acetone ≥98%
(2-Fluorophenyl)acetone
Formula: C₉H₉FO
MW: 152.17 g/mol
Boiling Pt: 47 °C
Density: 1.077
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000325
CAS Number: 2836-82-0
EINECS: 220-627-5