N-(3-Hydroxypropyl)morpholine 95%
Supplier: Thermo Scientific
Warning
Synonyms: 3-Morpholinopropanol, 3-Morpholino-1-propanol, 4-(3-Hydroxypropyl)morpholine, 3-(4-Morpholinyl)-1-propanol
H60376.03 H60376.06
H60376.03EA 50.6 EUR
H60376.03 H60376.06
N-(3-Hydroxypropyl)morpholine 95%
N-(3-Hydroxypropyl)morpholine
Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Boiling Pt: 240…241 °C
Density: 1.049
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD01713533
CAS Number: 4441-30-9