1,3,5-Triethylbenzene 95%
Supplier: Thermo Fisher Scientific
Warning
H60875.06 H60875.14
H60875.06EA 73.1 EUR
H60875.06 H60875.14
1,3,5-Triethylbenzene 95%
1,3,5-Triethylbenzene
Formula: C₁₂H₁₈
Boiling Pt: 215 °C
Melting Pt: 3 °C
Density: 0.862
Flash Pt: 76 °C (169 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009261
CAS Number: 102-25-0
EINECS: 203-017-3
UN: 3082
ADR: 9,III