2-Piperidinemethanol ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-(Hydroxymethyl)piperidine
H60178.09
H60178.18
H60178.09EA
95.4
EUR
H60178.09
H60178.18
2-Piperidinemethanol ≥97%
2-Piperidinemethanol
Formula:
C₆H₁₃NO Boiling Pt: 100…102 °C Melting Pt: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005987 CAS Number: 3433-37-2 EINECS: 222-333-2 |