4-Hydroxybenzenesulphonic acid 65% in water
Supplier: Thermo Fisher Scientific
Danger
Synonyms: p-Phenolsulphonic acid, Phenol-4-sulphonic acid, 4-Hydroxybenzenesulphonic acid hydrate
H59917.AK H59917.K2
H59917.AKEA 73.9 EUR
H59917.AK H59917.K2
4-Hydroxybenzenesulphonic acid 65% in water
4-Hydroxybenzenesulphonic acid
Formula: C₆H₆O₄S
Density: 1.337
Storage Temperature: Ambient
MDL Number: MFCD00673723
CAS Number: 98-67-9
EINECS: 202-691-6
UN: 1803
ADR: 8,II