3,3',5,5'-Tetramethylbenzidine dihydrochloride ≥99%
Supplier: Thermo Scientific
Warning
Synonyms: Dihydrochloride TMB, TMB-HCl, TMB 2HCl, TMB dihydrochloride
H60245.MD H60245.03
H60245.MDEA 57.3 EUR
H60245.MD H60245.03
3,3',5,5'-Tetramethylbenzidine dihydrochloride ≥99%
3,3',5,5'-Tetramethylbenzidine dihydrochloride
Formula: C₁₆H₂₀N₂·2HCl
MW: 313.27 g/mol
Melting Pt: 300 °C
Storage Temperature: Freezer
MDL Number: MFCD00012961
CAS Number: 64285-73-0
EINECS: 264-769-6