Reaction block systems for RH basic, RH digital, RCT basic, RET basic and RET control-visc magnetic hotplate stirrers
Supplier: IKA
442-0977EA 87.5 EUR
442-0977 442-0966 442-0967 442-0978 442-0968 442-0969 442-0984 442-0973 442-0962 IKAA25003280 442-0985 442-0974 442-0963 442-0975 442-0964 442-0976 442-0965 442-0981 442-0970 442-0982 IKAA20007954 442-0983
Reaction block systems for RH basic, RH digital, RCT basic, RET basic and RET control-visc magnetic hotplate stirrers
Block Heaters Blocks
Reaction blocks allow syntheses to be carried out in round-bottomed flasks at temperatures up to 180 °C, avoiding the use of oil baths. The system ensures optimum heat transfer from the heating plate directly into the medium. Uniform mixing is also guaranteed since there is no interference to the magnetic field from the aluminium blocks. Reaction blocks range from 100 to 5000 ml in size, and inserts accommodating round bottom flasks from 10 to 3000 ml are available. Due to the wide range of interchangeable inserts, this system can be used for a range of different applications with the RCT, RET and RH magnetic hotplate stirrers.
  • Perform safe and clean syntheses in round bottom flasks at temperatures up to 180 °C
  • Uniform mixing
  • Thermal conduction is directly into the reaction blocks and is very precise
  • PTFE coating prevents burning and helps ensure the system is safe to work with