Single channel pipettes, electronic, variable volume, Ovation®
Supplier: VISTALAB TECHNOLOGIES
1065-1250 1065-0250
VIST1065-1250EA 1420 EUR
VIST1065-1250 VIST1065-0250
Single channel pipettes, electronic, variable volume, Ovation®
Pipettes Micropipettes
These pipettes are the ideal ergonomic choice for users who need a comprehensive range of automated liquid handling functions. An easy-to-read display and simple-to-operate keypad allows fast recall and setup of user-defined pipetting protocols and selection of operating modes, volumes and speeds.
  • Fully electronic operation maximises pipetting efficiency for tasks like multiple dispensing, pipetting, reverse pipetting, mixing, serial diluting and automatic sequential dispensing
  • Colour-coded right and left-handed models are available
Delivery information: These models require a power supply unit (sold separately) to recharge the pipette's battery.