Packaging bags, MELAfol®
Supplier: Melag
MELAfol®
MELA00751EA 0 EUR
MELA00751
Packaging bags, MELAfol®
Bags Sterilisation Pouches
MELAfol® is a combination packaging solution. It is sealed against all germs, tear resistant, hot sealable, crease-resistant and easy to open (with a 'peelable' seal seam).
  • It has a handling indicator in accordance with EN 868-5 which colours from rose to brown upon sterilisation
  • The packaging can be opened through pulling apart the side seal sealing
  • Storage length in accordance with DIN 58953 part 7: up to 6 months