Occupational shoes, clogs, The Original, 7030/7031
Supplier: Abeba
7030/39 7030/40 7030/41 7030/46 7030/47 7031/37 7031/42 7031/35
ABEB7030/39EA 50.05 EUR
ABEB7030/39 ABEB7030/40 ABEB7030/41 ABEB7030/46 ABEB7030/47 114-8151 114-8156 114-8149
Occupational shoes, clogs, The Original, 7030/7031
Shoes and Boots
Anti-static clogs made from smooth or perforated leather upper and a sole with cross/circle profile resistant to animal fats, oil and fuel.
  • Leather insole
  • Sanitized® treatment reduces odour and bacterial growth
  • Foldable and adjustable heel strap
Colour: Black
Colour: Black
Certifikace: TÜV GS tested; EN ISO 20347:2012; OB, FO, SRC.