Cut resistant gloves, HyFlex® 70-205
Supplier: Ansell
70-205/7 70-205/8 70-205/9 70-205/10
112-3553EA 9.6 EUR
112-3553 112-3554 112-3555 112-3552
Cut resistant gloves, HyFlex® 70-205
Gloves
These knitted gloves are designed to provide superior mechanical strength in light to medium-duty handling and production applications. The Kevlar® reinforcement provides a very high cut resistance, offers enhanced comfort and dexterity.
  • Neptune® coating: glove fibres repel liquids, avoiding damage to the Kevlar® strands and extending glove life
  • Reinforced thumb welt increases abrasion resistance
  • Fully washable and can be reused many times
Ideal for applications involving metal and glass.
Certifikace: EN 388:2016 (134XB), EN 407 (X1XXXX); Cat. III
Order Now
SPECIFICATIONS
Model 70-205
Ambidextrous / hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Reusable
Liner material Kevlar®
Finish Neptune® coating
Gauge 13
Cuff style Knit wrist
EN norm EN 388:2016 (134XB), EN 407 (X1XXXX)