Cut resistant gloves, Camapur® Cut 627
Supplier: KCL by Honeywell
627.10 627.11 627.12 627.6 627.7 627.8 627.9
112-2976EA 374 EUR
112-2976 KCHL627.11 KCHL627.12 KCHL627.6 KCHL627.7 112-2977 112-2978
Cut resistant gloves, Camapur® Cut 627
Gloves
This glove made from HPPE fibres combines good sensitivity, mechanical resistance and high cut-resistance. The thin PU coating offers good sensitivity and prevents sweating.
  • Excellent grip, even when handling slightly oiled, wet and slippery items
  • Back of hand without coating for improved breathability
  • Dark grey colour for reduced susceptibility to dirt
  • Size of the glove is distinguishable at the knitted cuff through different colours
Ideal for assembly work with medium cutting risk, in sheet metal processing and the processing of plastic profiles.
Thickness: 1,2 ± 0,1 mm
Colour: grey/black
EN 388 (4341); Cat II
Certifikace: EN 388:2016 (4X41B); Cat. II