Measuring cylinders, conical
Supplier: CONTACTO
551/050 551/100 551/150 551/200
213-8020EA 35.4 EUR
213-8020 213-8021 213-8022 213-8023
Measuring cylinders, conical
Cylinders Measuring Cylinders
Stainless steel, with handle.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.