Blades for safety knives, No. 99
Supplier: Martor
233-0139EA 13.6 EUR
233-0139
Blades for safety knives, No. 99
Razors and Blades
Trapezoid blade for use with safety knives.
  • Deep-edged, two-sided ground edge
  • Double bevelled ground edge
  • Blade can be used twice
Mountable in:
SECUPRO MAXISAFE
SECUPRO MEGASAFE
SECUPRO 625
SECUNORM MULTISET
SECUNORM PROFI25