Lens cleaning box with spray and tissues
Supplier: MSA
111-1567EA 10.7 EUR
111-1567 111-1568
Lens cleaning box with spray and tissues
Cleaning Agents Lens Cleaning Solutions
Each box contains 110 ml of cleaning spray and a dispenser box with 280 lens cleaning wipes. The cleaning spray removes oil and grease spots from viewing panes, protective clothing, masks, goggles or cover spectacles quickly and easily. For an optimal cleaning result, use the spray in combination with the wipes.
  • Box can be mounted on the wall
  • Paper cleaning wipes are lint and silicone free
  • Wipes can be removed from the pull-out dispenser quickly and easily