Respiratory filters
Supplier: M S A
10097994 10108408 10115188 10115320 10097955 10097232
111-1557EA 29.4 EUR
111-1557 111-1559 111-1560 111-1562 111-1558 111-2538
Respiratory filters
Respirators Reusable Respirators Respirator Filters
Screw-in gas or combination filter for protection against toxic gases, vapours or dusts.
Certifikace: EN 148-1