Flasks, Erlenmeyer, with standard ground joint
Supplier: SMITH SCIENTIFIC
8SFE500/B19 8SFE150/B24 8SFE250/B19 8SFE100/B19
SMIT8SFE500/B19EA 2070 EUR
SMIT8SFE500/B19 SMIT8SFE150/B24 SMIT8SFE250/B19 SMIT8SFE100/B19
Flasks, Erlenmeyer, with standard ground joint
Flasks Erlenmeyer Flasks
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes