Capillary columns, TraceGOLD™ TG-17SilMS
Supplier: Thermo Fisher Scientific
26072-2750 26072-1430 26072-1540 26072-5780
555-8765EA 480 EUR
555-8765 555-8767 555-8766 555-8763
Capillary columns, TraceGOLD™ TG-17SilMS
These columns are ideal for the separation of active environmental compounds such as PAH's and pesticides.
  • TG-17SilMS fast increase speed of analysis by a factor of 3 to 10 times
  • High thermal stability - maximum temperatures up to 340/360 °C