Cut resistant gloves, Check & Go Green PU 5
Supplier: HONEYWELL SAFETY
2332545/8 2332545/10 2332545/7 2332545/9 2332545/11
112-4657EA 0 EUR
112-4657 112-4654 112-4656 112-4658 112-4655
Cut resistant gloves, Check & Go Green PU 5
Gloves
These knitted gloves in polyamide/HPPE fibres/elastane and glass fibres (outside) feature a green PU coating on the palm. Ideal for handling sharp parts in an oily or greasy environment.
  • Tight knit (gauge 13) gives the glove perfect support, limits penetration of dirt and offers a good sense of touch
  • Ventilated back optimises breathability
  • PU coating of the palm and fingertips provides outstanding abrasion resistance and oil protection
  • Composite fibre fully protected by polyamide increases the cut resistance
  • Colour-coding on top of the hand and coating colour helps safety manager and workers to visually recognise the level of cut protection
Typical application areas include: Automotive and part manufacturing, aviation building and construction, catering, energy or electricity, glass industries, metal and paper industries, logistics and maintenance.
Gauge: 13
Colour: White/green
EN 388 (4543); Cat. II
Certifikace: EN 388:2016 (3X43C); Cat. II