Glass density bottles, adjusted, MBL®
Supplier: SciLabware
BRE000 BRE004 BRE006
BIBBBRE000EA 139 EUR
BIBBBRE000 BIBBBRE004 BIBBBRE006
Glass density bottles, adjusted, MBL®
Specific Gravity Bottles
Borosilicate glass 3.3, Gay-Lussac type.
  • Adjusted ready for use - with exact volume engraved on the bottle
  • With individual capillary stoppers which are numbered and matched with the bottle
Complies with ISO 3507
Certifikace: ISO 3507